Assay Details
| Gene Symbol: BIVM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126520.1 | - | 1 | 2949 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BIVM-ERCC5 readthrough | BIVM-ERCC5 | ERCC5-202 | Sense | Chr. 13 - 102807146 - 102876001 on NCBI Build GRCh38 | |
| Downstream |
ERCC excision repair 5, endonuclease |
ERCC5 |
COFS3, ERCC5-201, ERCM2, UVDR, XPG, XPGC |
25.5kb | Sense | Chr. 13 - 102845841 - 102876001 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

BIVM
Genomic Map