Assay Details
Gene Symbol: NRXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105250.2 | NP_001098720.1 | 7 | 8221 | 81 | |
NM_001272020.1 | NP_001258949.1 | 6 | 8755 | 81 | ||
NM_004796.5 | NP_004787.2 | 17 | 9584 | 81 | ||
NM_138970.4 | NP_620426.2 | 6 | 8140 | 81 | ||
NR_073546.1 | 8 | 9533 | 81 | |||
NR_073547.1 | 21 | 12567 | 81 | |||
GenBank mRNA | AK126795.1 | - | 1 | 3547 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105370586 | LOC105370586 | -662.7kb | Sense | Chr. 14 - 79199982 - 79201632 on NCBI Build GRCh38 | |
Downstream |
deiodinase, iodothyronine, type II |
DIO2 |
5DII, D2, DIOII, SelY, TXDI2 |
329.3kb | Anti-sense | Chr. 14 - 80197525 - 80231054 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
