Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000514788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
ENST00000514788.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170367.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170367.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170367.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170367.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170367.hg.1 |
TC0500009998.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000514788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
ENST00000514788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009998.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170371.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170371.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170371.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170371.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170371.hg.1 |
TC0500009998.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500170363.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170363.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170363.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170363.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170363.hg.1 |
TC0500009998.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500170366.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170366.hg.1 |
ENST00000514788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170366.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170366.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170366.hg.1 |
ENST00000514788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170366.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170366.hg.1 |
TC0500009998.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500170368.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170368.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170368.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170368.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170368.hg.1 |
TC0500009998.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500170369.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170369.hg.1 |
ENST00000514788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170369.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170369.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170369.hg.1 |
ENST00000514788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170369.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170369.hg.1 |
TC0500009998.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500170365.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170365.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170365.hg.1 |
ENST00000503989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170365.hg.1 |
NR_103771.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170365.hg.1 |
TC0500009998.hg.1 |
Clariom D TC |