Assay Details
Gene Symbol: LOC100505625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103771.1 | 2 - 3 | 2758 | 63 | ||
GenBank mRNA | AK126802.1 | - | 2 - 3 | 2758 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | steroid 5 alpha-reductase 1 | SRD5A1 | S5AR 1 | -16.8kb | Anti-sense | Chr. 5 - 6633322 - 6669562 on NCBI Build GRCh38 |
Downstream | PAP associated domain containing 7 | PAPD7 |
LAK-1, LAK1, POLK, POLS, TRF4, TRF4-1, TRF41, TUTASE5 |
11.2kb | Anti-sense | Chr. 5 - 6713952 - 6757048 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908675 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742803 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742800 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908709 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908698 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742915 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742869 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742833 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908655 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742847 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742857 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742895 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908703 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742839 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908668 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742865 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908687 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908705 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908657 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742894 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908714 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908695 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742848 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908750 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908660 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742836 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908742 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742879 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908729 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742798 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742828 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908721 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742906 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908651 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908730 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742901 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908654 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742802 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908726 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908700 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742912 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742823 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908704 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742842 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742887 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908662 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742886 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908652 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908702 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15660770 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908672 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908706 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908699 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742878 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908693 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908708 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742829 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742815 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908712 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908685 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908718 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742826 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908717 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908669 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P03742891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908691 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908747 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908723 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908713 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908722 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P23908748 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 103665 | AF032110.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 103665 | AF238323.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 103665 | AK126001.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 103665 | BC051741.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 103665 | BX648746.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 14753 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 70799 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 26839 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 153895 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 477427 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 770664 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 568215 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 495016 | AK093200.1 | genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 680201 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 149874 | AB209685.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 149874 | AY692423.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 149874 | AY692424.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 149874 | BC010563.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 149874 | BC071654.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 149874 | CR749356.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 149874 | NM_197941.2 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 79546 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 92878 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 27628 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 30432 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 115065 | AL583915.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 115065 | BC003695.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 115065 | BC036911.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 115065 | NM_020964.2 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 359202 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 352747 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 331920 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 240101 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 313004 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 242840 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 93186 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 400270 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 411111 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 270159 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 29961 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 188986 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 177115 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 333758 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 124613 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P37077445 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P17018568 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P17018579 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P37077445 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P37077445 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P17018579 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 720 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17018572 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15077657 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15080610 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01173028 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17018579 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P37077445 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17018568 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01173024 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P17018579 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P37077445 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P17018568 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P17018572 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000514788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | ENST00000514788.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000514788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | ENST00000514788.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009998.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500170361.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500170361.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500170361.hg.1 | ENST00000503989.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500170361.hg.1 | NR_103771.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500170361.hg.1 | TC0500009998.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina human-6 v2.0 expression beadchip | ILMN_1864939 | - | - |
Illumina Sentrix Human-6 v2 Expression BeadChip | 1710673 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_110134 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1864939 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1864939 | - | - |
Illumina HumanWG-6_V2_0_R2 | ILMN_1864939 | - | - |
