Assay Details
| Gene Symbol: LOC100508227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130364.1 | - | 1 | 151 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
adiponectin, C1Q and collagen domain containing |
ADIPOQ |
ACDC, ACRP30, ADIPQTL1, ADPN, APM-1, APM1, GBP28 |
-74.1kb | Sense | Chr. 3 - 186842674 - 186858463 on NCBI Build GRCh38 |
| Downstream | ribosomal protein L39 like | RPL39L | RPL39L1 | 186.6kb | Anti-sense | Chr. 3 - 187120953 - 187139475 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100508227
Genomic Map