| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629357.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
ENST00000629576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007249.hg.1 |
NR_040085.1 |
Refseq |