Assay Details
| Gene Symbol: PTPN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK025546.1 | - | 1 | 1950 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 177 | TMEM177 | -301.0kb | Sense | Chr. 2 - 119679167 - 119682118 on NCBI Build GRCh38 | |
| Downstream | erythrocyte membrane protein band 4.1 like 5 | EPB41L5 |
BE37, YMO1, YRT |
27.6kb | Sense | Chr. 2 - 120013028 - 120179121 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PTPN4
Genomic Map