Assay Details
| Gene Symbol: WDFY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054833.1 | - | 1 | 2557 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4703 | MIR4703 | -36.7kb | Sense | Chr. 13 - 51552589 - 51552667 on NCBI Build GRCh38 | |
| Downstream | dehydrogenase/reductase 12 | DHRS12 | SDR40C1 | 175.4kb | Anti-sense | Chr. 13 - 51754856 - 51804206 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

WDFY2
Genomic Map