Assay Details
| Gene Symbol: OSER1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055067.1 | - | 1 | 2052 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | oxidative stress responsive serine rich 1 | OSER1 |
C20orf111, HSPC207, Osr1, Perit1, dJ1183I21.1 |
-119.0 | Anti-sense | Chr. 20 - 44195939 - 44217229 on NCBI Build GRCh38 |
| Downstream | ganglioside induced differentiation associated protein 1 like 1 | GDAP1L1 |
dJ881L22.1, dJ995J12.1.1 |
33.8kb | Sense | Chr. 20 - 44247099 - 44280917 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

OSER1-AS1
Genomic Map