Assay Details
Gene Symbol: LINC01441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110064.1 | 2 - 3 | 262 | 108 | ||
GenBank mRNA | AK056050.1 | - | 2 - 3 | 262 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1440 | LINC01440 | Anti-sense | Chr. 20 - 55423043 - 55426791 on NCBI Build GRCh38 | ||
Downstream | cerebellin 4 precursor | CBLN4 | CBLNL1 | 573.7kb | Sense | Chr. 20 - 55997357 - 56005472 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
