Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007822.hg.1 |
NR_038944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007822.hg.1 |
TC22000409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007823.hg.1 |
NR_038944.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22007823.hg.1 |
TC22000409.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200154872.hg.1 |
TC2200007736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200154872.hg.1 |
NR_038944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154872.hg.1 |
NR_038944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154870.hg.1 |
TC2200007736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200154870.hg.1 |
NR_038944.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200154870.hg.1 |
NR_038944.1 |
Refseq |