Assay Details
| Gene Symbol: RASEF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057300.1 | - | 1 | 1652 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SPATA31 subfamily D member 1 | SPATA31D1 | FAM75D1 | -1.1mb | Anti-sense | Chr. 9 - 81987404 - 81995256 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC102723989 | LOC102723989 | 170.0kb | Anti-sense | Chr. 9 - 83219288 - 83270833 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RASEF
Genomic Map