Assay Details
| Gene Symbol: LOC101928786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121582.1 | 2 - 3 | 527 | 60 | ||
| GenBank mRNA | AK126935.1 | - | 2 - 3 | 527 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105376331 | LOC105376331 | -93.5kb | Sense | Chr. 9 - 137943630 - 137947060 on NCBI Build GRCh38 | |
| Downstream | tubulin beta pseudogene 5 | TUBBP5 | 106.3kb | Anti-sense | Chr. 9 - 138150113 - 138177433 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928786
Genomic Map