Assay Details
| Gene Symbol: CMIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017023733.1 | XP_016879222.1 | 1 | 951 | 95 | |
| GenBank mRNA | AK127550.1 | - | 1 | 951 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6504 | MIR6504 | hsa-mir-6504 | -18.8kb | Sense | Chr. 16 - 81611348 - 81611408 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100129617 | LOC100129617 | 33.3kb | Sense | Chr. 16 - 81665354 - 81667276 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017023733.1 transcripts.
Set Membership: |
|

CMIP
Genomic Map