Assay Details
Gene Symbol: CRISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK128009.1 | - | 1 | 1749 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Rh-associated glycoprotein | RHAG |
CD241, OHS, OHST, RH2, RH50A, Rh50, Rh50GP, SLC42A1 |
-55.5kb | Sense | Chr. 6 - 49605175 - 49636874 on NCBI Build GRCh38 |
Downstream | cysteine rich secretory protein 3 | CRISP3 |
Aeg2, CRISP-3, CRS3, SGP28, dJ442L6.3 |
32.7kb | Sense | Chr. 6 - 49727376 - 49744455 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
