Assay Details
Gene Symbol: LOC101928780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK128544.1 | - | 3 - 4 | 3267 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TCF3 fusion partner | TFPT |
FB1, INO80F, amida |
-3.8kb | Sense | Chr. 19 - 54107013 - 54115675 on NCBI Build GRCh38 |
Downstream | CCR4-NOT transcription complex subunit 3 | CNOT3 |
LENG2, NOT3, NOT3H |
15.8kb | Anti-sense | Chr. 19 - 54137689 - 54155708 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
