Assay Details
Gene Symbol: PARVA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018222.4 | NP_060692.2 | 13 | 7797 | 77 | |
XM_005253015.3 | XP_005253072.1 | 13 | 7786 | 77 | ||
GenBank mRNA | AK128814.1 | - | 1 | 1149 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MICAL C-terminal like | MICALCL | Ebitein1 | -174.1kb | Sense | Chr. 11 - 12286900 - 12359144 on NCBI Build GRCh38 |
Downstream | TEA domain transcription factor 1 | TEAD1 |
AA, NTEF-1, REF1, TCF-13, TCF13, TEAD-1, TEF-1 |
139.1kb | Sense | Chr. 11 - 12674422 - 12944737 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
