Assay Details
Gene Symbol: LOC105379106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC062466.1 | - | 1 | 548 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 491 | LINC00491 | -310.5kb | Anti-sense | Chr. 5 - 102608492 - 102671464 on NCBI Build GRCh38 | |
Downstream | gypsy retrotransposon integrase 1 | GIN1 |
GIN-1, TGIN1, ZH2C2 |
102.5kb | Anti-sense | Chr. 5 - 103086000 - 103120149 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
