Assay Details
| Gene Symbol: MGST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC062473.1 | - | 1 | 745 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 15 member 5 | SLC15A5 | -70.0kb | Anti-sense | Chr. 12 - 16188485 - 16277685 on NCBI Build GRCh38 | |
| Downstream | LIM domain only 3 | LMO3 |
RBTN3, RBTNL2, RHOM3, Rhom-3 |
199.6kb | Anti-sense | Chr. 12 - 16548372 - 16610233 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MGST1
Genomic Map