Assay Details
| Gene Symbol: THPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB102892.1 | - | 1 | 53 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RNA polymerase II subunit H | POLR2H |
RPABC3, RPB17, RPB8 |
-7.3kb | Anti-sense | Chr. 3 - 184361710 - 184368595 on NCBI Build GRCh38 |
| Downstream | chordin | CHRD | 4.0kb | Anti-sense | Chr. 3 - 184380073 - 184389835 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

THPO
Genomic Map