Assay Details
| Gene Symbol: LOC105374587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK023940.1 | - | 1 | 2256 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | suppressor of cytokine signaling 5 | SOCS5 |
CIS6, CISH6, Cish5, SOCS-5 |
-15.0kb | Sense | Chr. 2 - 46698960 - 46762788 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1118 | LINC01118 | 36.4kb | Sense | Chr. 2 - 46816668 - 46822660 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105374587
Genomic Map