Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010877.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173930.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173930.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173930.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173930.hg.1 |
ENST00000427276.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600173930.hg.1 |
NR_108096.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173930.hg.1 |
NR_108097.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600173930.hg.1 |
TC0600010877.hg.1 |
Clariom D TC |