Assay Details
Gene Symbol: LOC401320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK097240.1 | - | 2 | 2251 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | gamma-glutamylcyclotransferase | GGCT |
C7orf24, CRF21, GCTG, GGC |
-55.1kb | Sense | Chr. 7 - 30496621 - 30504841 on NCBI Build GRCh38 |
Downstream | glycyl-tRNA synthetase | GARS |
CMT2D, DSMAV, GlyRS, HMN5, SMAD1 |
32.4kb | Anti-sense | Chr. 7 - 30594735 - 30634033 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
