Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152548.hg.1 |
TC2200007434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152548.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152548.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152548.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152548.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152548.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152548.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152545.hg.1 |
TC2200007434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152545.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152545.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152545.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152545.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152546.hg.1 |
TC2200007434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152546.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152546.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152546.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152546.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
ENST00000616875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007434.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152549.hg.1 |
TC2200007434.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200152549.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152549.hg.1 |
NR_109965.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200152549.hg.1 |
ENST00000417123.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200152549.hg.1 |
NR_109965.1 |
Refseq |