Assay Details
| Gene Symbol: RPAIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098491.1 | - | 1 | 49 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleoporin 88 | NUP88 | -5.4kb | Anti-sense | Chr. 17 - 5384833 - 5419739 on NCBI Build GRCh38 | |
| Downstream | complement C1q binding protein | C1QBP |
GC1QBP, HABP1, SF2p32, gC1Q-R, gC1qR, p32 |
7.3kb | Anti-sense | Chr. 17 - 5432779 - 5439151 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK098491.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000225698.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573204.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573406.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000573421.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000574444.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000575890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | ENST00000576122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012341.hg.1 | NM_001212.3 | Refseq |

RPAIN
Genomic Map