Assay Details
| Gene Symbol: ANKRD18CP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098764.1 | - | 1 | 546 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SUGT1-1300002K09Rik pseudogene | LOC100499484 | Anti-sense | Chr. 9 - 97238426 - 97297314 on NCBI Build GRCh38 | ||
| Downstream | coiled-coil domain containing 180 | CCDC180 |
BDAG1, C9orf174, KIAA1529 |
68.9kb | Anti-sense | Chr. 9 - 97307628 - 97377295 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ANKRD18CP
Genomic Map