Assay Details
| Gene Symbol: LL22NC01-81G9.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038954.1 | 1 | 47 | 98 | ||
| GenBank mRNA | AK130379.1 | - | 1 | 47 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | colony stimulating factor 2 receptor beta common subunit | CSF2RB |
CD131, CDw131, IL3RB, IL5RB, SMDP5 |
-24.8kb | Sense | Chr. 22 - 36913574 - 36940449 on NCBI Build GRCh38 |
| Downstream | testis expressed 33 | TEX33 |
C22orf33, EAN57, cE81G9.2 |
25.6kb | Anti-sense | Chr. 22 - 36991119 - 37007864 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000273.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK130379.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495323 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179013 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280775 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179050 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179049 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495345 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280782 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495332 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179003 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495339 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179029 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495363 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32178990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495368 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32178982 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32178991 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495317 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495352 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495391 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495316 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495328 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280799 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179001 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495314 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32178994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495322 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32178996 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495383 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179025 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495324 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495388 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495344 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495320 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179004 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280778 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495384 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179031 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495336 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495365 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179011 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32178971 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32178997 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495364 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495379 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17280781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495376 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32178977 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179016 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495362 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P32179002 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P11495321 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3364235 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43365 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3364235 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3364235 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03623328 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41523785 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22005010.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005010.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005010.hg.1 | TC22000273.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007275.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007275.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007275.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007275.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007275.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007275.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007275.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007275.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005008.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005008.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005008.hg.1 | TC22000273.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005009.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005009.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005009.hg.1 | TC22000273.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005011.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005011.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22005011.hg.1 | TC22000273.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200007275.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007275.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007275.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007275.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007275.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007275.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007275.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007275.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150968.hg.1 | TC2200007275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150968.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150968.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150968.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150968.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150965.hg.1 | TC2200007275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150965.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150965.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150965.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150965.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150966.hg.1 | TC2200007275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150966.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150966.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150966.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150966.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150967.hg.1 | TC2200007275.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200150967.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150967.hg.1 | NR_038954.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200150967.hg.1 | ENST00000400221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200150967.hg.1 | NR_038954.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020406 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020406 | - | - |

LL22NC01-81G9.3
Genomic Map