Assay Details
| Gene Symbol: RPL17P10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130593.1 | - | 1 | 153 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NADH:ubiquinone oxidoreductase subunit B3 | NDUFB3 |
B12, CI-B12 |
-16.6kb | Anti-sense | Chr. 2 - 201071739 - 201085750 on NCBI Build GRCh38 |
| Downstream | CASP8 and FADD like apoptosis regulator | CFLAR |
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS |
13.0kb | Anti-sense | Chr. 2 - 201116015 - 201172688 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK130593.1 transcripts.
This assay is in the 5'UTR region of the AK130593.1 transcripts.
Set Membership: |
|

RPL17P10
Genomic Map