Assay Details
| Gene Symbol: LOC105377146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135532.1 | 1 | 159 | 97 | ||
| GenBank mRNA | AK131014.1 | - | 1 | 149 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SUCLG2 antisense RNA 1 (head to head) | SUCLG2-AS1 | -552.9kb | Sense | Chr. 3 - 67654697 - 67947713 on NCBI Build GRCh38 | |
| Downstream |
family with sequence similarity 19 member A4, C-C motif chemokine like |
FAM19A4 |
TAFA-4, TAFA4 |
230.8kb | Anti-sense | Chr. 3 - 68731764 - 68932610 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105377146
Genomic Map