Assay Details
Gene Symbol: NADK2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC033402.1 | - | 2 | 447 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
NAD kinase 2, mitochondrial |
NADK2 |
C5orf33, DECRD, MNADK, NADKD1 |
Anti-sense | Chr. 5 - 36192589 - 36242279 on NCBI Build GRCh38 | |
Downstream | RAN binding protein 3 like | RANBP3L | 26.0kb | Anti-sense | Chr. 5 - 36246919 - 36301936 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
