Assay Details
| Gene Symbol: CTNNBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC035794.1 | - | 2 | 243 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calsyntenin 1 | CLSTN1 |
ALC-ALPHA, CDHR12, CST-1, CSTN1, PIK3CD, XB31alpha, alcalpha1, alcalpha2 |
-55.4kb | Sense | Chr. 1 - 9729021 - 9824526 on NCBI Build GRCh38 |
| Downstream | leucine zipper and CTNNBIP1 domain containing | LZIC | 41.2kb | Sense | Chr. 1 - 9922113 - 9943427 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CTNNBIP1
Genomic Map