Assay Details
Gene Symbol: LOC105370549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC039046.1 | - | 1 | 447 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105370550 | LOC105370550 | -37.2kb | Sense | Chr. 14 - 69183018 - 69260567 on NCBI Build GRCh38 | |
Downstream | enhancer of rudimentary homolog (Drosophila) | ERH | DROER | 81.5kb | Sense | Chr. 14 - 69380123 - 69398304 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
