Assay Details
| Gene Symbol: LOC101927346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC040672.2 | - | 2 | 554 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PDZRN3 antisense RNA 1 | PDZRN3-AS1 | -376.4kb | Sense | Chr. 3 - 73623568 - 73627899 on NCBI Build GRCh38 | |
| Downstream | contactin 3 | CNTN3 |
BIG-1, PANG, PCS |
257.0kb | Anti-sense | Chr. 3 - 74262568 - 74614448 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101927346
Genomic Map