Assay Details
| Gene Symbol: LOC101927790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC041445.1 | - | 2 | 646 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927766 | LOC101927766 | -65.2kb | Sense | Chr. 5 - 159776772 - 159871384 on NCBI Build GRCh38 | |
| Downstream | tetratricopeptide repeat domain 1 | TTC1 | TPR1 | 72.1kb | Anti-sense | Chr. 5 - 160009100 - 160065545 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101927790
Genomic Map