Assay Details
| Gene Symbol: LOC101928902 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134307.1 | 1 | 555 | 84 | ||
| GenBank mRNA | BC047297.1 | - | 1 | 555 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | solute carrier family 52 member 2 | SLC52A2 |
BVVLS2, D15Ertd747e, GPCR41, GPR172A, PAR1, RFT3, RFVT2, hRFT3 |
-10.6kb | Anti-sense | Chr. 8 - 144358547 - 144361286 on NCBI Build GRCh38 |
| Downstream | aarF domain containing kinase 5 | ADCK5 | 1.1kb | Anti-sense | Chr. 8 - 144373762 - 144393242 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101928902
Genomic Map