Assay Details
Gene Symbol: LINC00641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038971.1 | 1 | 48 | 89 | ||
GenBank mRNA | BX537733.1 | - | 1 | 48 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000914.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human CGH Microarray 4x180K | 165041 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 388641 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 326471 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 165020 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3543133 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P931030 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 29365 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48883 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02872029 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3543133 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3543133 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P931030 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P02872029 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P02872029 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P931030 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 14837 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 13883 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02872023 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19973854 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02872029 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02872020 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19973845 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P761678 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P346729 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P02872029 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P19973845 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 240902_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR14009889.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14009889.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14009889.hg.1 | TC14000914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14009892.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14009892.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14009892.hg.1 | TC14000914.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14009890.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14009890.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14009890.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14009890.hg.1 | TC14000914.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1400162187.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162187.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162187.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162187.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162187.hg.1 | TC1400008624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400162178.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162178.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162178.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162178.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162178.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162178.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162178.hg.1 | TC1400008624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008624.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162180.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162180.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162180.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162180.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162180.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162180.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162180.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162180.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162180.hg.1 | TC1400008624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400162181.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162181.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162181.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162181.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162181.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162181.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162181.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162181.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162181.hg.1 | TC1400008624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400162184.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162184.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162184.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162184.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162184.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162184.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162184.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162184.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162184.hg.1 | TC1400008624.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400162182.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162182.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162182.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162182.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162182.hg.1 | ENST00000555379.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162182.hg.1 | ENST00000555688.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400162182.hg.1 | NR_038970.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162182.hg.1 | NR_038971.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400162182.hg.1 | TC1400008624.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 240902_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 240902_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 240902_at6 | - | - |
Human Genome U95 Set | 62134_PM_at | - | - |
Human Genome U95 Set | 62134_at | - | - |
Human Genome U95 Set | 91262_at | - | - |
Human Genome U95 Set | 74355_PM_at | - | - |
Human Genome U95 Set | 91262_PM_at | - | - |
Human Genome U95 Set | 88306_PM_at | - | - |
Human Genome U95 Set | 88306_at | - | - |
Human Genome U95 Set | 74355_at | - | - |
Human Genome U133 Set | 240902_at | - | - |
Human Genome U133 Set | 240902_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050569 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050568 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050569 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050568 | - | - |
