Assay Details
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124690.1 | - | 1 | 3350 | 111 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ZBED5 antisense RNA 1 | ZBED5-AS1 | -391.6kb | Anti-sense | Chr. 11 - 10858217 - 10879276 on NCBI Build GRCh38 | |
Downstream | casein kinase 2 alpha 3 | CSNK2A3 | CSNK2A1P | 77.3kb | Sense | Chr. 11 - 11351942 - 11353357 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
