Assay Details
| Gene Symbol: UGT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128174.2 | NP_001121646.1 | 6 | 3980 | 83 | |
| NM_001322112.1 | NP_001309041.1 | 7 | 4083 | 83 | ||
| NM_001322113.1 | NP_001309042.1 | 6 | 4063 | 83 | ||
| NM_001322114.1 | NP_001309043.1 | 6 | 3909 | 83 | ||
| NM_003360.4 | NP_003351.2 | 5 | 3919 | 83 | ||
| GenBank mRNA | AL137342.1 | - | 1 | 1450 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 577 | MIR577 |
MIRN577, hsa-mir-577, mir-577 |
-19.8kb | Sense | Chr. 4 - 114656759 - 114656854 on NCBI Build GRCh38 |
| Downstream | N-deacetylase and N-sulfotransferase 4 | NDST4 |
N-HSST, N-HSST 4, NDST-4, NHSST4 |
149.5kb | Anti-sense | Chr. 4 - 114824122 - 115114098 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

UGT8
Genomic Map