Assay Details
| Gene Symbol: ZNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC017945.1 | - | 1 | 1246 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 107 | ZNF107 |
Y8, ZFD25, ZNF588, smap-7 |
-141.0kb | Sense | Chr. 7 - 64665012 - 64711582 on NCBI Build GRCh38 |
| Downstream | eukaryotic translation elongation factor 1 delta pseudogene 4 | EEF1DP4 | 8.9kb | Sense | Chr. 7 - 64862951 - 64864450 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZNF138
Genomic Map