Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013142.hg.1 |
TC03000725.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03013142.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205343.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205343.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205343.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300205343.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300205343.hg.1 |
TC0300013874.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000466490.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000481356.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000484684.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
ENST00000494297.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013874.hg.1 |
NM_021203.3 |
Refseq |