Assay Details
| Gene Symbol: PAN3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029383.1 | 2 | 1989 | 80 | ||
| GenBank mRNA | BC045730.1 | - | 1 | 1150 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PAN3 poly(A) specific ribonuclease subunit | PAN3 | Anti-sense | Chr. 13 - 28138079 - 28295338 on NCBI Build GRCh38 | ||
| Downstream | fms related tyrosine kinase 1 | FLT1 |
FLT, FLT-1, VEGFR-1, VEGFR1 |
162.2kb | Sense | Chr. 13 - 28300346 - 28495128 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000516.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188413 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741799 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741838 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188397 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188362 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188402 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188395 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188358 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188354 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188393 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188353 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741817 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188372 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741815 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188415 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188394 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188408 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188423 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741821 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08741850 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188369 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188400 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188350 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188420 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29188409 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 369526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188412 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188414 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188413 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188400 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188360 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741850 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741820 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188404 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16621656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188415 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188394 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188417 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188353 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188355 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741833 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741850 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188413 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188412 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188415 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188417 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16621656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741804 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188400 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741838 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188394 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188360 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188414 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741843 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188355 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188362 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188404 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08741820 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29188353 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02743948 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P414040 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 243092_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005830.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005830.hg.1 | TC13000516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005829.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13005829.hg.1 | TC13000516.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008454.hg.1 | ENST00000563843.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008454.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008454.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008454.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157005.hg.1 | ENST00000563843.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157005.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157005.hg.1 | ENST00000563843.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300157005.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157005.hg.1 | TC1300008454.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300157008.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157008.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157008.hg.1 | TC1300008454.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008454.hg.1 | ENST00000563843.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008454.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008454.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008454.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157006.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157006.hg.1 | NR_029383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300157006.hg.1 | TC1300008454.hg.1 | Clariom D TC |
| Human Genome U95 Set | 58110_PM_at | - | - |
| Human Genome U95 Set | 58110_at | - | - |
| Human Genome U133 Set | 243092_PM_at | - | - |
| Human Genome U133 Set | 243092_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049362 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A049362 | - | - |

PAN3-AS1
Genomic Map