Assay Details
| Gene Symbol: DCLRE1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033855.2 | NP_001029027.1 | 14 | 5531 | 108 | |
| NM_001033857.2 | NP_001029029.1 | 15 | 5616 | 108 | ||
| NM_001033858.2 | NP_001029030.1 | 16 | 5938 | 108 | ||
| NM_001289076.1 | NP_001276005.1 | 12 | 5390 | 108 | ||
| NM_001289077.1 | NP_001276006.1 | 15 | 5662 | 108 | ||
| NM_001289078.1 | NP_001276007.1 | 12 | 5423 | 108 | ||
| NM_001289079.1 | NP_001276008.1 | 16 | 5984 | 108 | ||
| NM_022487.3 | NP_071932.2 | 13 | 5475 | 108 | ||
| NR_110297.1 | 17 | 5884 | 108 | |||
| GenBank mRNA | BC067233.1 | - | 1 | 455 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | suppressor of variegation 3-9 homolog 2 | SUV39H2 | KMT1B | -296.0 | Anti-sense | Chr. 10 - 14878783 - 14904315 on NCBI Build GRCh38 |
| Downstream | meiosis/spermiogenesis associated 1 | MEIG1 |
SPATA39, bA2K17.3 |
48.4kb | Anti-sense | Chr. 10 - 14959439 - 14972851 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

DCLRE1C
Genomic Map