Assay Details
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC073842.1 | - | 1 | 449 | 74 | |
BI821381.1 | - | 1 | 448 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein S (alpha) | PROS1 |
PROS, PS21, PS22, PS23, PS24, PS25, PSA, THPH5, THPH6 |
-6.0kb | Anti-sense | Chr. 3 - 93873037 - 93974090 on NCBI Build GRCh38 |
Downstream | syntaxin 19 | STX19 | 32.5kb | Anti-sense | Chr. 3 - 94014365 - 94029281 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
