Assay Details
| Gene Symbol: ITPKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017001211.1 | XP_016856700.1 | 3 | 9661 | 71 | |
| GenBank mRNA | BC086865.1 | - | 1 | 2151 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ITPKB intronic transcript 1 | ITPKB-IT1 | -46.7kb | Sense | Chr. 1 - 226656080 - 226675068 on NCBI Build GRCh38 | |
| Downstream | presenilin 2 | PSEN2 |
AD3L, AD4, CMD1V, PS2, STM2 |
146.2kb | Anti-sense | Chr. 1 - 226870572 - 226903829 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ITPKB
Genomic Map