Assay Details
| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005249807.4 | XP_005249864.1 | 7 | 9679 | 174 | |
| XM_006715752.2 | XP_006715815.1 | 8 | 9647 | 174 | ||
| XM_017012414.1 | XP_016867903.1 | 7 | 9606 | 174 | ||
| XM_017012415.1 | XP_016867904.1 | 8 | 9708 | 174 | ||
| XM_017012417.1 | XP_016867906.1 | 8 | 8110 | 174 | ||
| XM_017012418.1 | XP_016867907.1 | 7 | 7980 | 174 | ||
| XM_017012421.1 | XP_016867910.1 | 8 | 9830 | 174 | ||
| XM_017012422.1 | XP_016867911.1 | 8 | 9719 | 174 | ||
| XM_017012424.1 | XP_016867913.1 | 7 | 9622 | 174 | ||
| XM_017012430.1 | XP_016867919.1 | 8 | 8082 | 174 | ||
| GenBank mRNA | BX537586.1 | - | 1 | 3143 | 174 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 |
HECW1 | NEDL1 | -60.7kb | Anti-sense | Chr. 7 - 43112580 - 43566001 on NCBI Build GRCh38 |
| Downstream | biliverdin reductase A | BLVRA |
BLVR, BVR, BVRA |
130.6kb | Anti-sense | Chr. 7 - 43758153 - 43807342 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

COA1
Genomic Map