Assay Details
| Gene Symbol: HPRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000194.2 | NP_000185.1 | 3 | 343 | 64 | |
| GenBank mRNA | AK313435.1 | - | 3 | 323 | 64 | |
| BC000578.2 | - | 3 | 215 | 3163726 | 64 | |
| BT019350.1 | - | 3 | 176 | 64 | ||
| CR407645.1 | - | 3 | 176 | 64 | ||
| KR817914.1 | - | 2 | 59 | 64 | ||
| L29382.1 | - | 3 | 176 | 64 | ||
| M24772.1 | - | 160 | 64 | |||
| M31642.1 | - | 3 | 261 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

HPRT1
Genomic Map