Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022796.hg.1 |
ENST00000268957.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022796.hg.1 |
NM_001243877.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022796.hg.1 |
TC17001698.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022797.hg.1 |
ENST00000268957.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022797.hg.1 |
NM_001243877.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022797.hg.1 |
TC17001698.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700189509.hg.1 |
ENST00000268957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189509.hg.1 |
NM_001243877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189509.hg.1 |
ENST00000268957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189509.hg.1 |
NM_001243877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189509.hg.1 |
TC1700011121.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700189510.hg.1 |
ENST00000268957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189510.hg.1 |
NM_001243877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189510.hg.1 |
ENST00000268957.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189510.hg.1 |
NM_001243877.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189510.hg.1 |
TC1700011121.hg.1 |
Clariom D TC |