Assay Details
Gene Symbol: BHLHA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164405.1 | NP_001157877.1 | 1 | 251 | 93 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000010.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3372688 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3245606 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 50628 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57035 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3245606 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3372688 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3245606 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3372688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189356 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737928 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737920 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737931 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189359 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189352 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737939 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189370 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189349 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737929 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737924 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189363 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189371 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737927 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189373 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737934 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189366 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189356 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737920 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737934 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737939 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189370 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189359 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737928 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189349 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189366 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737929 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737924 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737931 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189371 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189363 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189373 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30737927 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10189352 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.1 ST Array | 8003633 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1700006499.hg.1 | ENST00000391429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006499.hg.1 | ENST00000391429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700006499.hg.1 | NM_001164405.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17000058.hg.1 | ENST00000391429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17000058.hg.1 | NM_001164405.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17000058.hg.1 | TC17000010.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700144368.hg.1 | ENST00000391429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700144368.hg.1 | NM_001164405.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700144368.hg.1 | ENST00000391429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700144368.hg.1 | NM_001164405.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700144368.hg.1 | TC1700006499.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700006499.hg.1 | ENST00000391429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006499.hg.1 | ENST00000391429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700006499.hg.1 | NM_001164405.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008944 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A008944 | - | - |
