Assay Details
| Gene Symbol: PDE4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104631.1 | NP_001098101.1 | 8 - 9 | 1365 | 88 | |
| NM_001165899.1 | NP_001159371.1 | 10 - 11 | 1256 | 88 | ||
| NM_001197218.1 | NP_001184147.1 | 8 - 9 | 1104 | 88 | ||
| NM_001197219.1 | NP_001184148.1 | 8 - 9 | 882 | 88 | ||
| NM_001197220.1 | NP_001184149.1 | 8 - 9 | 1520 | 88 | ||
| NM_001197221.1 | NP_001184150.1 | 4 - 5 | 840 | 88 | ||
| NM_001197222.1 | NP_001184151.1 | 3 - 4 | 926 | 88 | ||
| NM_001197223.1 | NP_001184152.1 | 3 - 4 | 716 | 88 | ||
| NM_006203.4 | NP_006194.2 | 8 - 9 | 908 | 88 | ||
| XM_005248537.3 | XP_005248594.1 | 8 - 9 | 2062 | 88 | ||
| XM_005248538.4 | XP_005248595.1 | 9 - 10 | 3963 | 88 | ||
| XM_011543469.1 | XP_011541771.1 | 11 - 12 | 1345 | 88 | ||
| XM_011543470.2 | XP_011541772.1 | 11 - 12 | 1269 | 88 | ||
| XM_011543471.1 | XP_011541773.1 | 10 - 11 | 1222 | 88 | ||
| XM_011543473.1 | XP_011541775.1 | 10 - 11 | 1170 | 88 | ||
| XM_011543477.2 | XP_011541779.1 | 10 - 11 | 1172 | 88 | ||
| XM_017009565.1 | XP_016865054.1 | 11 - 12 | 1382 | 88 | ||
| XM_017009566.1 | XP_016865055.1 | 11 - 12 | 1272 | 88 | ||
| XM_017009567.1 | XP_016865056.1 | 9 - 10 | 1010 | 88 | ||
| XM_017009568.1 | XP_016865057.1 | 11 - 12 | 1256 | 88 | ||
| XM_017009569.1 | XP_016865058.1 | 10 - 11 | 1135 | 88 | ||
| XM_017009570.1 | XP_016865059.1 | 7 - 8 | 1955 | 88 | ||
| XM_017009571.1 | XP_016865060.1 | 10 - 11 | 1178 | 88 | ||
| GenBank mRNA | AF012073.1 | - | 8 - 9 | 1118 | 88 | |
| AF012074.1 | - | 4 - 5 | 854 | 88 | ||
| AF536975.1 | - | 3 - 4 | 523 | 88 | ||
| AF536976.1 | - | 10 - 11 | 1079 | 88 | ||
| AF536977.1 | - | 8 - 9 | 882 | 88 | ||
| AJ250852.1 | - | 4 - 5 | 450 | 88 | ||
| AJ250854.1 | - | 9 - 10 | 1045 | 88 | ||
| AY245866.1 | - | 10 - 11 | 1221 | 88 | ||
| AY245867.1 | - | 8 - 9 | 1520 | 88 | ||
| AY388960.1 | - | 8 - 9 | 803 | 88 | ||
| BC036319.1 | - | 3 - 4 | 718 | 4828512 | 88 | |
| L20970.1 | - | 8 - 9 | 916 | 88 | ||
| U02882.1 | - | 7 - 8 | 686 | 88 | ||
| U50157.1 | - | 3 - 4 | 524 | 88 | ||
| U50158.1 | - | 4 - 5 | 438 | 88 | ||
| U50159.1 | - | 8 - 9 | 859 | 88 | ||
| U79571.1 | - | 3 - 4 | 929 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001389.hg.1
Set Membership: |
|

PDE4D
Genomic Map