Assay Details
| Gene Symbol: HMGCL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166059.1 | NP_001159531.1 | 4 - 5 | 394 | 68 | |
| GenBank mRNA | AK300733.1 | - | 4 - 5 | 363 | 68 | |
| FJ472654.1 | - | 2 - 3 | 173 | 68 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
liver development
leucine catabolic process
acyl-CoA metabolic process
mitochondrion organization
response to nutrient
response to starvation
ketone body biosynthetic process
protein tetramerization
response to fatty acid
fatty-acyl-CoA binding
magnesium ion binding
hydroxymethylglutaryl-CoA lyase activity
receptor binding
manganese ion binding
carboxylic acid binding
protein homodimerization activity
metal ion binding
liver development
leucine catabolic process
acyl-CoA metabolic process
mitochondrion organization
response to nutrient
response to starvation
ketone body biosynthetic process
protein tetramerization
response to fatty acid
fatty-acyl-CoA binding
magnesium ion binding
hydroxymethylglutaryl-CoA lyase activity
receptor binding
manganese ion binding
carboxylic acid binding
protein homodimerization activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000479458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000479458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000479458.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000498698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000498698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000498698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000235958.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000374490.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000436439.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000479458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000479458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000479458.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000496907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000498698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000498698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000498698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000509389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | ENST00000513148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_000191.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013303.hg.1 | NM_001166059.1 | Refseq |

HMGCL
Genomic Map